Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
56 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609882 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MTF2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
MTF2 - metal response element binding transcription factor 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001164391.1 | Intron | NP_001157863.1 | ||||
NM_001164392.1 | Intron | NP_001157864.1 | ||||
NM_001164393.1 | Intron | NP_001157865.1 | ||||
NM_007358.3 | Intron | NP_031384.1 | ||||
XM_011541016.1 | Intron | XP_011539318.1 | ||||
XM_011541017.2 | Intron | XP_011539319.1 | ||||
XM_011541019.1 | Intron | XP_011539321.1 | ||||
XM_017000676.1 | Intron | XP_016856165.1 | ||||
XM_017000677.1 | Intron | XP_016856166.1 | ||||
XM_017000678.1 | Intron | XP_016856167.1 | ||||
XM_017000679.1 | Intron | XP_016856168.1 |
Set Membership: |
HapMap |