Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCTGGGGAGTGGGTCCCTGGCTTA[C/T]GGACACAGTCTACCCCCATCCCGTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603399 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
KCNK15-AS1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
KCNK15-AS1 - KCNK15 antisense RNA 1 (head to head) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
WISP2 - WNT1 inducible signaling pathway protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001323369.1 | Intron | NP_001310298.1 | ||||
NM_001323370.1 | Intron | NP_001310299.1 | ||||
NM_003881.3 | Intron | NP_003872.1 | ||||
XM_017028116.1 | Intron | XP_016883605.1 | ||||
XM_017028117.1 | Intron | XP_016883606.1 |
Set Membership: |
HapMap Validated |