Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCTGCTGCCGGGTATGTGCACGGC[A/G]GCTGGGGGAGCCCTGCGACCAACTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603399 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
KCNK15-AS1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
KCNK15-AS1 - KCNK15 antisense RNA 1 (head to head) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
WISP2 - WNT1 inducible signaling pathway protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001323369.1 | 371 | Missense Mutation | CAG,CGG | Q,R 59 | NP_001310298.1 | |
NM_001323370.1 | 371 | Missense Mutation | CAG,CGG | Q,R 59 | NP_001310299.1 | |
NM_003881.3 | 371 | Missense Mutation | CAG,CGG | Q,R 59 | NP_003872.1 | |
XM_017028116.1 | 371 | UTR 5 | XP_016883605.1 | |||
XM_017028117.1 | 371 | Missense Mutation | CAG,CGG | Q,R 117 | XP_016883606.1 |