Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTATTTGTTGTAGCAACATTACCAG[A/C]TGACTAGTATTACTACCATTACTTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF559 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
ZNF559 - zinc finger protein 559 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001202406.1 | Intron | NP_001189335.1 | ||||
NM_001202407.1 | Intron | NP_001189336.1 | ||||
NM_001202408.1 | Intron | NP_001189337.1 | ||||
NM_001202409.1 | Intron | NP_001189338.1 | ||||
NM_001202410.1 | Intron | NP_001189339.1 | ||||
NM_001202411.1 | Intron | NP_001189340.1 | ||||
NM_001202412.1 | Intron | NP_001189341.1 | ||||
NM_032497.2 | Intron | NP_115886.1 |
ZNF559-ZNF177 - ZNF559-ZNF177 readthrough | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001172650.2 | Intron | NP_001166121.1 | ||||
NM_001202425.1 | Intron | NP_001189354.1 |
Set Membership: |
HapMap Validated |