Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCTTAAATGAAAGGGTGGCAATAT[C/T]TTCAGGCATTCCCAGTGGTTGCCTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
TCP11L1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
TCP11L1 - t-complex 11 like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145541.1 | Intron | NP_001139013.1 | ||||
NM_018393.3 | Intron | NP_060863.3 | ||||
XM_011520204.2 | Intron | XP_011518506.1 | ||||
XM_011520205.2 | Intron | XP_011518507.1 | ||||
XM_011520206.2 | Intron | XP_011518508.1 | ||||
XM_011520207.2 | Intron | XP_011518509.1 | ||||
XM_017017989.1 | Intron | XP_016873478.1 | ||||
XM_017017990.1 | Intron | XP_016873479.1 | ||||
XM_017017991.1 | Intron | XP_016873480.1 | ||||
XM_017017992.1 | Intron | XP_016873481.1 |
Set Membership: |
HapMap |