Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AACGAGAGGGCAAAAGGGGAGACAG[G/T]GAGTATAAATAACTTCCATGGAGTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
OVCH1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
OVCH1 - ovochymase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_183378.2 | Intron | NP_899234.2 | ||||
XM_011520638.2 | Intron | XP_011518940.1 | ||||
XM_011520639.2 | Intron | XP_011518941.1 | ||||
XM_011520640.2 | Intron | XP_011518942.1 | ||||
XM_011520641.2 | Intron | XP_011518943.1 | ||||
XM_011520642.2 | Intron | XP_011518944.1 | ||||
XM_017019256.1 | Intron | XP_016874745.1 | ||||
XM_017019257.1 | Intron | XP_016874746.1 |
OVCH1-AS1 - OVCH1 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |