Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616150 | ||||||||||||||||||||||||||||||||
Literature Links: |
SLC25A48 PubMed Links | ||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU)
|
|||
EAS - Not Available | African American - Not Available | YRI (Yoruba)
|
|||
SAS - Not Available | Chinese - Not Available | JPT (Japanese)
|
|||
AFR - Not Available | Japanese - Not Available | CHB (Han Chinese)
|
|||
EUR - Not Available | |||||
AMR - Not Available |
SLC25A48 - solute carrier family 25 member 48 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_145282.4 | Intron | NP_660325.4 | ||||
XM_005271903.1 | Intron | XP_005271960.1 | ||||
XM_006714544.1 | Intron | XP_006714607.1 | ||||
XM_006714546.1 | Intron | XP_006714609.1 | ||||
XM_006714550.1 | Intron | XP_006714613.1 | ||||
XM_006714552.2 | Intron | XP_006714615.1 | ||||
XM_011543196.1 | Intron | XP_011541498.1 | ||||
XM_011543199.1 | Intron | XP_011541501.1 | ||||
XM_017009086.1 | Intron | XP_016864575.1 | ||||
XM_017009087.1 | Intron | XP_016864576.1 | ||||
XM_017009088.1 | Intron | XP_016864577.1 | ||||
XM_017009089.1 | Intron | XP_016864578.1 | ||||
XM_017009090.1 | Intron | XP_016864579.1 | ||||
XM_017009091.1 | Intron | XP_016864580.1 | ||||
XM_017009092.1 | Intron | XP_016864581.1 |
Set Membership: |
HapMap |