Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGGATTAGGGCTTAGCTCATAATA[C/T]TGGAATCAGGACACAGCCTATGACA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616432 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ARHGEF18 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ARHGEF18 - Rho/Rac guanine nucleotide exchange factor 18 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001130955.1 | Intron | NP_001124427.1 | ||||
NM_015318.3 | Intron | NP_056133.2 | ||||
XM_005272464.4 | Intron | XP_005272521.1 | ||||
XM_006722705.3 | Intron | XP_006722768.1 | ||||
XM_006722706.3 | Intron | XP_006722769.1 | ||||
XM_006722708.2 | Intron | XP_006722771.1 | ||||
XM_011527835.2 | Intron | XP_011526137.1 | ||||
XM_011527836.2 | Intron | XP_011526138.1 | ||||
XM_011527837.2 | Intron | XP_011526139.1 | ||||
XM_011527838.2 | Intron | XP_011526140.1 | ||||
XM_011527839.2 | Intron | XP_011526141.1 | ||||
XM_011527840.1 | Intron | XP_011526142.1 | ||||
XM_011527841.2 | Intron | XP_011526143.1 |