Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTACAATAGCCACACACAAATAATA[C/T]CTGTTAGGTTGGTGCAAAAGTAATT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610291 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SV2C PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
SV2C - synaptic vesicle glycoprotein 2C | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001297716.1 | Intron | NP_001284645.1 | ||||
NM_014979.3 | Intron | NP_055794.3 | ||||
XM_011543281.2 | Intron | XP_011541583.1 | ||||
XM_011543282.2 | Intron | XP_011541584.2 | ||||
XM_017009244.1 | Intron | XP_016864733.1 |