Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 608074 | |||||||||||||||||||||||
Literature Links: |
PDCD6IP PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
PDCD6IP - programmed cell death 6 interacting protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001162429.2 | Intron | NP_001155901.1 | ||||
NM_001256192.1 | Intron | NP_001243121.1 | ||||
NM_013374.5 | Intron | NP_037506.2 | ||||
XM_011533252.1 | Intron | XP_011531554.1 | ||||
XM_011533253.1 | Intron | XP_011531555.1 | ||||
XM_017005488.1 | Intron | XP_016860977.1 |