Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGATTCTGACCCTTCCTAAACCAC[G/T]CCTAAGATCAGTGCTTGAGATATTT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602977 MIM: 191845 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
GP2 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
GP2 - glycoprotein 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
UMOD - uromodulin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001008389.2 | Intron | NP_001008390.1 | ||||
NM_001278614.1 | Intron | NP_001265543.1 | ||||
NM_003361.3 | Intron | NP_003352.2 | ||||
XM_011545934.1 | Intron | XP_011544236.1 | ||||
XM_011545935.1 | Intron | XP_011544237.1 | ||||
XM_011545936.1 | Intron | XP_011544238.1 | ||||
XM_011545937.1 | Intron | XP_011544239.1 | ||||
XM_011545938.1 | Intron | XP_011544240.1 | ||||
XM_011545940.1 | Intron | XP_011544242.1 | ||||
XM_017023641.1 | Intron | XP_016879130.1 |
Set Membership: |
HapMap |