Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCAGGCTCCTCTTCAGGGTACAAC[C/G]TTCCACCACCTTTCCACATACACGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607442 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
EML4 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
EML4 - echinoderm microtubule associated protein like 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145076.2 | Intron | NP_001138548.1 | ||||
NM_019063.4 | Intron | NP_061936.2 | ||||
XM_005264267.1 | Intron | XP_005264324.1 | ||||
XM_005264268.3 | Intron | XP_005264325.1 | ||||
XM_006711991.1 | Intron | XP_006712054.1 | ||||
XM_006711992.2 | Intron | XP_006712055.1 |
LOC102723824 - uncharacterized LOC102723824 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |