Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGTGAGCCACTGCGCCTGGTCTAA[C/T]GTGACATATTTTGCTAACCATTGAG
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600580 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CLCN3 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese)
|
||||||
EUR
|
||||||||
AMR
|
CLCN3 - chloride voltage-gated channel 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001243372.1 | Intron | NP_001230301.1 | ||||
NM_001243374.1 | Intron | NP_001230303.1 | ||||
NM_001829.3 | Intron | NP_001820.2 | ||||
NM_173872.3 | Intron | NP_776297.2 | ||||
XM_005262726.2 | Intron | XP_005262783.1 | ||||
XM_011531586.2 | Intron | XP_011529888.1 |
Set Membership: |
HapMap |