Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTATGGATTTTGTAATATTACTCA[A/G]AGTATCATAAGACTCTGCTTATTTC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 603283 | ||||||||||||||||||||
Literature Links: |
TRDN PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TRDN - triadin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001251987.1 | Intron | NP_001238916.1 | ||||
NM_001256020.1 | Intron | NP_001242949.1 | ||||
NM_001256021.1 | Intron | NP_001242950.1 | ||||
NM_001256022.1 | Intron | NP_001242951.1 | ||||
NM_006073.3 | Intron | NP_006064.2 |