Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
68 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603949 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
RAB29 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
RAB29 - RAB29, member RAS oncogene family | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001135662.1 | 1777 | UTR 3 | NP_001129134.1 | |||
NM_001135663.1 | 1777 | UTR 3 | NP_001129135.1 | |||
NM_001135664.1 | 1777 | UTR 3 | NP_001129136.1 | |||
NM_003929.2 | 1777 | UTR 3 | NP_003920.1 | |||
XM_005245569.1 | 1777 | UTR 3 | XP_005245626.1 | |||
XM_005245570.1 | 1777 | UTR 3 | XP_005245627.1 | |||
XM_005245571.1 | 1777 | UTR 3 | XP_005245628.1 | |||
XM_006711605.3 | 1777 | UTR 3 | XP_006711668.1 | |||
XM_006711606.2 | 1777 | UTR 3 | XP_006711669.1 | |||
XM_017002748.1 | 1777 | UTR 3 | XP_016858237.1 | |||
XM_017002749.1 | 1777 | UTR 3 | XP_016858238.1 | |||
XM_017002750.1 | 1777 | UTR 3 | XP_016858239.1 |
Set Membership: |
HapMap |