Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607467 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CLECL1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global - Not Available | Caucasian
|
CEPH (CEU)
|
||||||
EAS - Not Available | African American
|
YRI (Yoruba)
|
||||||
SAS - Not Available | Japanese
|
JPT (Japanese)
|
||||||
AFR - Not Available | Chinese
|
CHB (Han Chinese)
|
||||||
EUR - Not Available | ||||||||
AMR - Not Available |
CLECL1 - C-type lectin like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001253750.1 | 623 | UTR 3 | NP_001240679.1 | |||
NM_001267701.1 | 623 | UTR 3 | NP_001254630.1 | |||
NM_172004.3 | 623 | Intron | NP_742001.1 | |||
XM_011520574.2 | 623 | Intron | XP_011518876.1 | |||
XM_017018885.1 | 623 | Intron | XP_016874374.1 |
Set Membership: |
HapMap Validated |