Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGACACTGGCCGGGGGTGGGGCTCG[A/C]AAGGCATAGTGCTGCAGGATGACAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
BREA2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
BREA2 - breast cancer estrogen-induced apoptosis 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZNF707 - zinc finger protein 707 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001100598.1 | Intron | NP_001094068.1 | ||||
NM_001100599.1 | Intron | NP_001094069.1 | ||||
NM_001288805.1 | Intron | NP_001275734.1 | ||||
NM_001288806.1 | Intron | NP_001275735.1 | ||||
NM_001288807.1 | Intron | NP_001275736.1 | ||||
NM_001288808.1 | Intron | NP_001275737.1 | ||||
NM_001288809.1 | Intron | NP_001275738.1 | ||||
NM_173831.3 | Intron | NP_776192.2 | ||||
XM_005250883.3 | Intron | XP_005250940.1 | ||||
XM_006716542.3 | Intron | XP_006716605.1 | ||||
XM_006716543.3 | Intron | XP_006716606.1 | ||||
XM_011516977.2 | Intron | XP_011515279.1 | ||||
XM_011516979.2 | Intron | XP_011515281.1 | ||||
XM_017013335.1 | Intron | XP_016868824.1 |
Set Membership: |
HapMap |