Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTTGAGGGGCCCCCGGGCCACGAC[G/A]GTGGCCGCCCGGGGCCGGTTCAGCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 176911 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PRKAR1B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PRKAR1B - protein kinase cAMP-dependent type I regulatory subunit beta | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001164758.1 | 1058 | Silent Mutation | ACC,ACT | T,T 338 | NP_001158230.1 | |
NM_001164759.1 | 1058 | Silent Mutation | ACC,ACT | T,T 338 | NP_001158231.1 | |
NM_001164760.1 | 1058 | Silent Mutation | ACC,ACT | T,T 338 | NP_001158232.1 | |
NM_001164761.1 | 1058 | Silent Mutation | ACC,ACT | T,T 338 | NP_001158233.1 | |
NM_001164762.1 | 1058 | Silent Mutation | ACC,ACT | T,T 338 | NP_001158234.1 | |
NM_002735.2 | 1058 | Silent Mutation | ACC,ACT | T,T 338 | NP_002726.1 | |
XM_011515446.2 | 1058 | Silent Mutation | ACC,ACT | T,T 283 | XP_011513748.1 | |
XM_017012433.1 | 1058 | Silent Mutation | ACC,ACT | T,T 208 | XP_016867922.1 | |
XM_017012434.1 | 1058 | Silent Mutation | ACC,ACT | T,T 186 | XP_016867923.1 |