Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TACTTACAGGCTGTAAAAGCTTCTC[A/C]AGTCTAAAAATATCTGAAAACTAAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
26 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613513 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZC3H11A PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ZC3H11A - zinc finger CCCH-type containing 11A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001319238.1 | Intron | NP_001306167.1 | ||||
NM_001319239.1 | Intron | NP_001306168.1 | ||||
NM_014827.5 | Intron | NP_055642.3 | ||||
XM_005245643.2 | Intron | XP_005245700.1 | ||||
XM_005245644.3 | Intron | XP_005245701.1 | ||||
XM_006711673.2 | Intron | XP_006711736.1 | ||||
XM_011510199.1 | Intron | XP_011508501.1 | ||||
XM_011510200.1 | Intron | XP_011508502.1 | ||||
XM_011510201.1 | Intron | XP_011508503.1 | ||||
XM_011510202.1 | Intron | XP_011508504.1 | ||||
XM_017002956.1 | Intron | XP_016858445.1 | ||||
XM_017002957.1 | Intron | XP_016858446.1 | ||||
XM_017002958.1 | Intron | XP_016858447.1 | ||||
XM_017002959.1 | Intron | XP_016858448.1 | ||||
XM_017002960.1 | Intron | XP_016858449.1 | ||||
XM_017002961.1 | Intron | XP_016858450.1 | ||||
XM_017002962.1 | Intron | XP_016858451.1 | ||||
XM_017002963.1 | Intron | XP_016858452.1 | ||||
XM_017002964.1 | Intron | XP_016858453.1 | ||||
XM_017002965.1 | Intron | XP_016858454.1 |
Set Membership: |
HapMap |