Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCACTTTGACATATTGTCTGGGAGT[A/G]CTTACTGTAGGGTTTGGGATGGAAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611236 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
TMEM38B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TMEM38B - transmembrane protein 38B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018112.2 | Intron | NP_060582.1 | ||||
XM_005252075.2 | Intron | XP_005252132.1 | ||||
XM_005252076.2 | Intron | XP_005252133.1 | ||||
XM_005252077.3 | Intron | XP_005252134.1 | ||||
XM_011518829.1 | Intron | XP_011517131.1 | ||||
XM_011518831.2 | Intron | XP_011517133.1 | ||||
XM_011518832.2 | Intron | XP_011517134.1 | ||||
XM_011518833.2 | Intron | XP_011517135.1 |