Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611877 MIM: 615628 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
BAIAP2L1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
BAIAP2L1 - BAI1 associated protein 2 like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018842.4 | 3099 | UTR 3 | NP_061330.2 |
BRI3 - brain protein I3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001159491.1 | 3099 | Intron | NP_001152963.1 | |||
NM_015379.4 | 3099 | Intron | NP_056194.1 | |||
XM_017011931.1 | 3099 | Intron | XP_016867420.1 | |||
XM_017011932.1 | 3099 | Intron | XP_016867421.1 | |||
XM_017011933.1 | 3099 | Intron | XP_016867422.1 | |||
XM_017011934.1 | 3099 | Intron | XP_016867423.1 | |||
XM_017011935.1 | 3099 | Intron | XP_016867424.1 | |||
XM_017011936.1 | 3099 | Intron | XP_016867425.1 |
Set Membership: |
HapMap |