Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTGCTGCTCTGGAATATTCTCTCTC[A/G]CGCACATGCGCTCTTTGTGTGCGTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ENDOV PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU) - Not Available | ||||||
EAS
|
African American
|
YRI (Yoruba) - Not Available | ||||||
SAS
|
Japanese
|
CHB (Han Chinese) - Not Available | ||||||
AFR
|
Chinese
|
JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
LOC100294362 - uncharacterized LOC100294362 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR4730 - microRNA 4730 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |