Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CATTTATGATTCCATTTTTCCCACT[A/G]ACTGTAGGTTCTCAAGAGTAGGGTG
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ENDOV PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese)
|
||||||
EUR
|
||||||||
AMR
|
LOC100294362 - uncharacterized LOC100294362 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR4730 - microRNA 4730 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |