Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGAGAAATGTGGACAGATTTCAGAG[A/G]TATTTCAGATTTAATGACTCCAAAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 143089 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
FOXN2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
FOXN2 - forkhead box N2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002158.3 | Intron | NP_002149.2 | ||||
XM_005264282.2 | Intron | XP_005264339.1 | ||||
XM_005264283.2 | Intron | XP_005264340.1 | ||||
XM_006712001.3 | Intron | XP_006712064.1 | ||||
XM_006712002.3 | Intron | XP_006712065.1 | ||||
XM_011532808.2 | Intron | XP_011531110.1 | ||||
XM_017003956.1 | Intron | XP_016859445.1 | ||||
XM_017003957.1 | Intron | XP_016859446.1 | ||||
XM_017003958.1 | Intron | XP_016859447.1 | ||||
XM_017003959.1 | Intron | XP_016859448.1 |