Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAAAAATAGGGAAAAAACCCAAGAT[A/G]AGTGAGATACATTCCAATATGCTTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604450 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PSMD12 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PSMD12 - proteasome 26S subunit, non-ATPase 12 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001316341.1 | Intron | NP_001303270.1 | ||||
NM_002816.4 | Intron | NP_002807.1 | ||||
NM_174871.3 | Intron | NP_777360.1 |