Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610327 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
RUFY1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
RUFY1 - RUN and FYVE domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001040451.2 | Intron | NP_001035541.1 | ||||
NM_001040452.2 | Intron | NP_001035542.1 | ||||
NM_025158.4 | Intron | NP_079434.3 | ||||
XM_005265993.4 | Intron | XP_005266050.1 | ||||
XM_006714921.3 | Intron | XP_006714984.2 | ||||
XM_006714922.3 | Intron | XP_006714985.1 | ||||
XM_017009891.1 | Intron | XP_016865380.1 | ||||
XM_017009892.1 | Intron | XP_016865381.1 | ||||
XM_017009893.1 | Intron | XP_016865382.1 | ||||
XM_017009894.1 | Intron | XP_016865383.1 | ||||
XM_017009895.1 | Intron | XP_016865384.1 |