Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTCGTGAGTGGCACACGTAGGGCAA[C/T]TCGATTGCTCTGCGTGCGGAATCGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
25 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600773 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
RNU11 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
RNU11 - RNA, U11 small nuclear | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TAF12 - TATA-box binding protein associated factor 12 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001135218.1 | 70 | Intron | NP_001128690.1 | |||
NM_005644.3 | 70 | Intron | NP_005635.1 | |||
XM_017002186.1 | 70 | UTR 5 | XP_016857675.1 |