Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGGCTGTAGATCAGAGGATTCAAC[A/G]TGGGAATCACCACTGTGTAGAACAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608492 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
OR5F1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
OR5F1 - olfactory receptor family 5 subfamily F member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003697.1 | 851 | Missense Mutation | ACG,ATG | T,M 284 | NP_003688.1 |
OR7E5P - olfactory receptor family 7 subfamily E member 5 pseudogene | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |