Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTTCTACTGGGAAAGATGGAGTGG[A/G]GGATGCATGGGCAGGACCCGCCATG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 171500 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ACP1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ACP1 - acid phosphatase 1, soluble | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
FAM150B - family with sequence similarity 150 member B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001002919.2 | Intron | NP_001002919.2 | ||||
XM_005264689.4 | Intron | XP_005264746.1 | ||||
XM_006711879.3 | Intron | XP_006711942.1 | ||||
XM_011510342.2 | Intron | XP_011508644.1 |
LOC101927262 - melanoma-associated antigen C1-like | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |