Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600844 MIM: 174762 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
P2RX2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
|||||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
P2RX2 - purinergic receptor P2X 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282164.1 | 7605 | Intron | NP_001269093.1 | |||
NM_001282165.1 | 7605 | Intron | NP_001269094.1 | |||
NM_012226.4 | 7605 | Intron | NP_036358.2 | |||
NM_016318.3 | 7605 | Intron | NP_057402.1 | |||
NM_170682.3 | 7605 | Intron | NP_733782.1 | |||
NM_170683.3 | 7605 | Intron | NP_733783.1 | |||
NM_174872.2 | 7605 | Intron | NP_777361.1 | |||
NM_174873.2 | 7605 | Intron | NP_777362.1 | |||
XM_005266154.3 | 7605 | Intron | XP_005266211.1 | |||
XM_005266155.4 | 7605 | Intron | XP_005266212.1 | |||
XM_005266156.4 | 7605 | Intron | XP_005266213.1 | |||
XM_011534786.2 | 7605 | Intron | XP_011533088.1 | |||
XM_017019035.1 | 7605 | Intron | XP_016874524.1 |
POLE - DNA polymerase epsilon, catalytic subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006231.3 | 7605 | UTR 3 | NP_006222.2 | |||
XM_011534795.2 | 7605 | Intron | XP_011533097.1 | |||
XM_011534797.2 | 7605 | Intron | XP_011533099.1 | |||
XM_011534798.1 | 7605 | Intron | XP_011533100.1 | |||
XM_011534799.2 | 7605 | Intron | XP_011533101.1 | |||
XM_011534800.2 | 7605 | Intron | XP_011533102.1 | |||
XM_011534802.2 | 7605 | Intron | XP_011533104.1 |
Set Membership: |
HapMap Validated |