Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAAACCCCTGTCCAAGGAGGGCAGC[G/A]ATCTTCTCCAACTGCCTGGGGGGAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604568 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DOC2B PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU) - Not Available | ||||||
EAS
|
African American
|
YRI (Yoruba) - Not Available | ||||||
SAS
|
Japanese
|
CHB (Han Chinese) - Not Available | ||||||
AFR
|
Chinese
|
JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
DOC2B - double C2 domain beta | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003585.4 | Intron | NP_003576.2 | ||||
XM_011535363.2 | Intron | XP_011533665.1 | ||||
XM_017025227.1 | Intron | XP_016880716.1 |
LOC100506371 - uncharacterized LOC100506371 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
Validated |