Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611449 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
EEF1AKMT1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
EEF1AKMT1 - eukaryotic translation elongation factor 1 alpha lysine methyltransferase 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
XPO4 - exportin 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_022459.4 | 4867 | UTR 3 | NP_071904.4 | |||
XM_005266502.1 | 4867 | UTR 3 | XP_005266559.1 | |||
XM_005266503.3 | 4867 | UTR 3 | XP_005266560.1 | |||
XM_005266504.3 | 4867 | UTR 3 | XP_005266561.1 | |||
XM_011535197.1 | 4867 | UTR 3 | XP_011533499.1 | |||
XM_011535198.1 | 4867 | UTR 3 | XP_011533500.1 | |||
XM_011535199.1 | 4867 | UTR 3 | XP_011533501.1 | |||
XM_011535200.2 | 4867 | UTR 3 | XP_011533502.1 | |||
XM_017020704.1 | 4867 | UTR 3 | XP_016876193.1 | |||
XM_017020705.1 | 4867 | UTR 3 | XP_016876194.1 | |||
XM_017020706.1 | 4867 | UTR 3 | XP_016876195.1 |
Set Membership: |
HapMap |