Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTTGGGCTTCTTAGATCCTTTCGC[C/T]GGCAAGACACTGTCTACAGTCTGAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606410 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ANTXR1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ANTXR1 - anthrax toxin receptor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018153.3 | Intron | NP_060623.2 | ||||
NM_032208.2 | Intron | NP_115584.1 | ||||
NM_053034.2 | Intron | NP_444262.1 | ||||
XM_017005075.1 | Intron | XP_016860564.1 | ||||
XM_017005076.1 | Intron | XP_016860565.1 | ||||
XM_017005077.1 | Intron | XP_016860566.1 |