Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTGGTGTGGCCTCCGCCTGGGTGT[A/G]TCCTGGTTAATTCCTGTGCCTACTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611061 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
FAM20C PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
FAM20C - family with sequence similarity 20 member C | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020223.3 | 535 | Intron | NP_064608.2 | |||
XM_017012450.1 | 535 | Intron | XP_016867939.1 | |||
XM_017012451.1 | 535 | Intron | XP_016867940.1 | |||
XM_017012452.1 | 535 | Intron | XP_016867941.1 | |||
XM_017012453.1 | 535 | Intron | XP_016867942.1 | |||
XM_017012454.1 | 535 | Intron | XP_016867943.1 | |||
XM_017012455.1 | 535 | UTR 5 | XP_016867944.1 |