Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607879 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
EXOC1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
|||||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
EXOC1 - exocyst complex component 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001024924.1 | Intron | NP_001020095.1 | ||||
NM_018261.3 | Intron | NP_060731.2 | ||||
NM_178237.2 | Intron | NP_839955.1 | ||||
XM_005265747.2 | Intron | XP_005265804.1 | ||||
XM_005265748.1 | Intron | XP_005265805.1 | ||||
XM_005265750.2 | Intron | XP_005265807.1 | ||||
XM_017008405.1 | Intron | XP_016863894.1 | ||||
XM_017008406.1 | Intron | XP_016863895.1 | ||||
XM_017008407.1 | Intron | XP_016863896.1 | ||||
XM_017008408.1 | Intron | XP_016863897.1 | ||||
XM_017008409.1 | Intron | XP_016863898.1 |
Set Membership: |
HapMap Validated |