Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607695 MIM: 603449 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
EEFSEC PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
EEFSEC - eukaryotic elongation factor, selenocysteine-tRNA specific | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_021937.4 | Intron | NP_068756.2 | ||||
XM_005247696.2 | Intron | XP_005247753.1 | ||||
XM_011513066.2 | Intron | XP_011511368.1 | ||||
XM_017006975.1 | Intron | XP_016862464.1 | ||||
XM_017006976.1 | Intron | XP_016862465.1 | ||||
XM_017006977.1 | Intron | XP_016862466.1 | ||||
XM_017006978.1 | Intron | XP_016862467.1 | ||||
XM_017006979.1 | Intron | XP_016862468.1 | ||||
XM_017006980.1 | Intron | XP_016862469.1 | ||||
XM_017006981.1 | Intron | XP_016862470.1 |
RUVBL1 - RuvB like AAA ATPase 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |