Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTGTTAGAAGATCACATGAGCTGCT[A/C]TTGATACAATTAATTGGAAAAGGGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610292 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
BANK1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
BANK1 - B-cell scaffold protein with ankyrin repeats 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001083907.2 | Intron | NP_001077376.2 | ||||
NM_001127507.2 | Intron | NP_001120979.2 | ||||
NM_017935.4 | Intron | NP_060405.4 | ||||
XM_017008337.1 | Intron | XP_016863826.1 |