Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613316 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
WDFY4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
WDFY4 - WDFY family member 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020945.1 | Intron | NP_065996.1 | ||||
XM_005270004.3 | Intron | XP_005270061.1 | ||||
XM_011539986.2 | Intron | XP_011538288.2 | ||||
XM_011539987.2 | Intron | XP_011538289.2 | ||||
XM_011539988.2 | Intron | XP_011538290.1 | ||||
XM_011539990.2 | Intron | XP_011538292.1 | ||||
XM_011539991.2 | Intron | XP_011538293.1 | ||||
XM_011539992.2 | Intron | XP_011538294.1 | ||||
XM_017016463.1 | Intron | XP_016871952.1 | ||||
XM_017016464.1 | Intron | XP_016871953.1 | ||||
XM_017016465.1 | Intron | XP_016871954.1 |