Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTCCCTGTCTTGAAGCAGAACACA[C/T]CTCCCTTGAGTTTCTCCAACTAGCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602070 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
NRP2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NRP2 - neuropilin 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003872.2 | Intron | NP_003863.2 | ||||
NM_018534.3 | Intron | NP_061004.3 | ||||
NM_201264.1 | Intron | NP_957716.1 | ||||
NM_201266.1 | Intron | NP_957718.1 | ||||
NM_201267.1 | Intron | NP_957719.1 | ||||
NM_201279.1 | Intron | NP_958436.1 | ||||
XM_005246933.3 | Intron | XP_005246990.2 | ||||
XM_005246934.3 | Intron | XP_005246991.2 | ||||
XM_017005185.1 | Intron | XP_016860674.1 | ||||
XM_017005186.1 | Intron | XP_016860675.1 | ||||
XM_017005187.1 | Intron | XP_016860676.1 | ||||
XM_017005188.1 | Intron | XP_016860677.1 |