Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 147370 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
IGF1R PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
IGF1R - insulin like growth factor 1 receptor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000875.4 | Intron | NP_000866.1 | ||||
NM_001291858.1 | Intron | NP_001278787.1 | ||||
XM_011521516.2 | Intron | XP_011519818.1 | ||||
XM_011521517.2 | Intron | XP_011519819.1 | ||||
XM_017022136.1 | Intron | XP_016877625.1 | ||||
XM_017022137.1 | Intron | XP_016877626.1 | ||||
XM_017022138.1 | Intron | XP_016877627.1 | ||||
XM_017022139.1 | Intron | XP_016877628.1 |
IRAIN - IGF1R antisense imprinted non-protein coding RNA | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap Validated |