Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATCCTCAGACTGGGACTTCTGTGTA[A/G]TTCTGCACCTCCCAGCCATAAAACT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605182 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC100507747 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
LOC100507747 - uncharacterized LOC100507747 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RASA3 - RAS p21 protein activator 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001320821.1 | 3383 | UTR 3 | NP_001307750.1 | |||
NM_001320822.1 | 3383 | UTR 3 | NP_001307751.1 | |||
NM_007368.3 | 3383 | UTR 3 | NP_031394.2 | |||
XM_011534841.2 | 3383 | UTR 3 | XP_011533143.1 | |||
XM_011534842.2 | 3383 | UTR 3 | XP_011533144.1 | |||
XM_017020438.1 | 3383 | UTR 3 | XP_016875927.1 | |||
XM_017020439.1 | 3383 | UTR 3 | XP_016875928.1 |
Set Membership: |
HapMap |