Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTTAGTACCAAATTGATGCAGAGAG[C/G]AAGGAAGACAAGGGGGTAGGAGCAT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
PAGE2B PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PAGE2B - PAGE family member 2B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001015038.2 | Intron | NP_001015038.1 | ||||
XM_011530785.2 | Intron | XP_011529087.1 | ||||
XM_011530786.2 | Intron | XP_011529088.1 | ||||
XM_011530787.2 | Intron | XP_011529089.1 | ||||
XM_017029513.1 | Intron | XP_016885002.1 |