Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCAGCAGCGAGCTGGACGTGAACA[C/T]CGACATCTACTCCAAGGTGCTGGTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 162651 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC107987285 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LOC107987285 - uncharacterized LOC107987285 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NTSR1 - neurotensin receptor 1 (high affinity) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002531.2 | 544 | Missense Mutation | ACC,ATC | T,I 58 | NP_002522.2 | |
XM_011528827.2 | 544 | Missense Mutation | ACC,ATC | T,I 58 | XP_011527129.1 |