Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AATCTCTGTAAGGACTCTATGGGCT[A/G]GAAAAGTTTTAAAATAAATTTATTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
TBC1D8B PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
TBC1D8B - TBC1 domain family member 8B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_017752.2 | Intron | NP_060222.2 | ||||
NM_198881.1 | Intron | NP_942582.1 | ||||
XM_006724669.1 | Intron | XP_006724732.1 | ||||
XM_006724670.3 | Intron | XP_006724733.1 |
Set Membership: |
HapMap |