Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
TMEM108 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
TMEM108 - transmembrane protein 108 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001136469.2 | Intron | NP_001129941.1 | ||||
NM_001282865.1 | Intron | NP_001269794.1 | ||||
NM_023943.3 | Intron | NP_076432.1 | ||||
XM_011513097.2 | Intron | XP_011511399.1 | ||||
XM_011513098.2 | Intron | XP_011511400.1 | ||||
XM_017007080.1 | Intron | XP_016862569.1 | ||||
XM_017007081.1 | Intron | XP_016862570.1 | ||||
XM_017007082.1 | Intron | XP_016862571.1 |