Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTGACCATGGGAATTTTGATCCCGC[C/T]CCTACTAAGCCTGAAATGAAGGACA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603804 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
NEURL1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
NEURL1 - neuralized E3 ubiquitin protein ligase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004210.4 | Intron | NP_004201.3 | ||||
XM_005270269.3 | Intron | XP_005270326.1 | ||||
XM_005270270.3 | Intron | XP_005270327.1 | ||||
XM_011540331.2 | Intron | XP_011538633.1 | ||||
XM_011540332.2 | Intron | XP_011538634.1 | ||||
XM_011540333.2 | Intron | XP_011538635.1 | ||||
XM_011540335.2 | Intron | XP_011538637.1 | ||||
XM_017016909.1 | Intron | XP_016872398.1 |
NEURL1-AS1 - NEURL1 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |