Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGGAGGCAATGACAGGTATGTATC[A/T]TATGAGATCAGAAGTTCTTTGTGAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609770 MIM: 611707 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
JAML PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
JAML - junction adhesion molecule like | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MPZL3 - myelin protein zero like 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286152.1 | Intron | NP_001273081.1 | ||||
NM_198275.2 | Intron | NP_938016.1 | ||||
XM_006718775.2 | Intron | XP_006718838.1 | ||||
XM_017017304.1 | Intron | XP_016872793.1 |
Set Membership: |
HapMap |