Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCTTGTTGTATTTAAGCTTGGTAC[A/G]CGTCTTAGAATAAATAAACCAATGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601291 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
UGT8 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU) - Not Available | ||||||
EAS
|
African American
|
YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
UGT8 - UDP glycosyltransferase 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001128174.2 | Intron | NP_001121646.1 | ||||
NM_001322112.1 | Intron | NP_001309041.1 | ||||
NM_001322113.1 | Intron | NP_001309042.1 | ||||
NM_001322114.1 | Intron | NP_001309043.1 | ||||
NM_003360.4 | Intron | NP_003351.2 |