Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTGGCATGTTCAAAGAGATCTCATG[A/G]CAAGAGAGTAAGGAAGAGAGAGAAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605010 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SPINK14 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
SPINK14 - serine peptidase inhibitor, Kazal type 14 (putative) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001001325.2 | 78 | Intron | NP_001001325.1 | |||
XM_017009466.1 | 78 | Intron | XP_016864955.1 | |||
XM_017009467.1 | 78 | UTR 5 | XP_016864956.1 | |||
XM_017009468.1 | 78 | UTR 5 | XP_016864957.1 | |||
XM_017009469.1 | 78 | Intron | XP_016864958.1 |
SPINK5 - serine peptidase inhibitor, Kazal type 5 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |