Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCTCCCCACCTGAGTTGAGAGTAC[A/G]TAGGCCTTTATCTCTGCACCCATCC
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604762 | |||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SWAP70 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese)
|
|||||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||||||||
EUR
|
|||||||||||
AMR
|
SWAP70 - SWAP switching B-cell complex 70kDa subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001297714.1 | Intron | NP_001284643.1 | ||||
NM_015055.3 | Intron | NP_055870.2 | ||||
XM_005252830.4 | Intron | XP_005252887.1 |
Set Membership: |
HapMap Validated |